The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013891.4 | NP_038919.1 | 1 - 2 | 400 | 162 |
GenBank mRNA | AB019436.1 | - | 1 - 2 | 336 | 161 |
AK078863.1 | - | 1 - 2 | 401 | 162 | |
BC012648.1 | - | 1 - 2 | 401 | 158 |