Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ppm1j
Entrez Gene ID
71887
Gene Name
protein phosphatase 1J
Gene Aliases
2310008J22Rik, PP2Czeta, Ppp2cz
Chromosome Location
Chr.3: 104781006 - 104786019 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027982.2 NP_082258.2 3 - 4 762 117
GenBank mRNA AK009235.1 - 3 - 4 701 117
AY184802.1 - 3 - 4 762 117
BC117498.1 - 3 - 4 758 117
BC121788.1 - 3 - 4 757 117

View Full Product Details