Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mov10
Entrez Gene ID
17454
Gene Name
Moloney leukemia virus 10
Gene Aliases
C77703, Mov-10
Chromosome Location
Chr.3: 104794828 - 104818599 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163440.1 NP_001156912.1 7 - 8 1399 71
NM_001163441.1 NP_001156913.1 7 - 8 1275 71
NM_008619.2 NP_032645.2 6 - 7 1376 71
GenBank mRNA AK004542.1 - 7 - 8 1250 71
AK150204.1 - 7 - 8 1259 71
AK159295.1 - 7 - 8 1411 71
AK169206.1 - 7 - 8 1370 71
AK173224.1 - 6 - 7 1289 71
BC053743.1 - 6 - 7 1295 71
X52574.1 - 6 - 7 1313 71

View Full Product Details