Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc2a13
Entrez Gene ID
239606
Gene Name
solute carrier family 2 (facilitated glucose transporter), member 13
Gene Aliases
6530403A04, A630029G22Rik, AI505012, Gm308
Chromosome Location
Chr.15: 91267691 - 91573261 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033633.3 NP_001028805.2 7 - 8 1573 98
GenBank mRNA AK147341.1 - 7 - 8 1574 98
BC119586.1 - 8 - 9 1140 98
BC119587.1 - 7 - 8 1082 98

View Full Product Details