The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033633.3 | NP_001028805.2 | 9 - 10 | 1856 | 82 |
GenBank mRNA | AK147341.1 | - | 9 - 10 | 1857 | 82 |
BC119586.1 | - | 10 - 11 | 1423 | 82 | |
BC119587.1 | - | 9 - 10 | 1365 | 82 |