The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_197987.2 | NP_932104.1 | 5 - 6 | 768 | 60 |
GenBank mRNA | AB093271.1 | - | 5 - 6 | 753 | 60 |
AK140822.1 | - | 5 - 6 | 769 | 60 | |
BC058678.1 | - | 5 - 6 | 766 | 60 | |
BC059070.1 | - | 5 - 6 | 734 | 60 | |
BC078416.1 | - | 5 - 6 | 754 | 60 |