Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ylpm1
Entrez Gene ID
56531
Gene Name
YLP motif containing 1
Gene Aliases
A930013E17Rik, ZAP, Zap3
Chromosome Location
Chr.12: 84996285 - 85074533 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178363.3 NP_848140.3 9 - 10 5376 98
GenBank mRNA AB033168.1 - 9 - 10 3049 98
AK162250.1 - 1 - 2 85 98

View Full Product Details