The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030017.3 | NP_084293.1 | 6 - 7 | 973 | 63 |
GenBank mRNA | AK020927.1 | - | 6 - 7 | 970 | 63 |
BC016204.1 | - | 6 - 7 | 920 | 63 | |
BC067213.1 | - | 5 - 6 | 2094 | 63 |