Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Luc7l
Entrez Gene ID
66978
Gene Name
Luc7 homolog (S. cerevisiae)-like
Gene Aliases
1810045C04Rik, 2410018D03Rik
Chromosome Location
Chr.17: 26245364 - 26285510 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028190.3 NP_082466.2 9 - 10 1092 68
GenBank mRNA AK017655.1 - 9 - 10 1052 68
CF951283.1 - 5 - 6 488 68

View Full Product Details