The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139152.1 | NP_631891.1 | 5 - 6 | 1219 | 62 |
GenBank mRNA | AY057054.1 | - | 5 - 6 | 1219 | 62 |
BC107370.1 | - | 3 - 4 | 588 | 62 | |
BC107371.1 | - | 4 - 5 | 427 | 62 | |
BC127152.1 | - | 3 - 4 | 588 | 62 | |
BC141295.1 | - | 4 - 5 | 1185 | 62 | |
BC145621.1 | - | 4 - 5 | 1185 | 62 |