Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Samm50
Entrez Gene ID
68653
Gene Name
sorting and assembly machinery component 50 homolog (S. cerevisiae)
Gene Aliases
1110030L07Rik
Chromosome Location
Chr.15: 84192233 - 84214303 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178614.4 NP_848729.1 14 - 15 1494 80
GenBank mRNA AK003990.1 - 14 - 15 1449 80
AK039949.1 - 14 - 15 1494 80
AK041783.1 - - 1488 80
AK084395.1 - 14 - 15 1488 80
AK086633.1 - - 1490 80
AK144975.1 - 14 - 15 80
AK159740.1 - 14 - 15 1506 80
AK161378.1 - 14 - 15 1484 80
AK168510.1 - 14 - 15 1450 80
AK169066.1 - 14 - 15 1484 80
AK169351.1 - 14 - 15 1451 80
BC119178.1 - 14 - 15 1488 80
BC119180.1 - 14 - 15 1488 80

View Full Product Details