Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Samm50
Entrez Gene ID
68653
Gene Name
sorting and assembly machinery component 50 homolog (S. cerevisiae)
Gene Aliases
1110030L07Rik
Chromosome Location
Chr.15: 84192233 - 84214303 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178614.4 NP_848729.1 5 - 6 561 83
GenBank mRNA AK003990.1 - 5 - 6 517 83
AK039949.1 - 5 - 6 561 83
AK041783.1 - - 555 83
AK084395.1 - 5 - 6 555 83
AK086633.1 - - 553 83
AK144975.1 - 5 - 6 562 83
AK159740.1 - 5 - 6 573 83
AK161378.1 - 5 - 6 551 83
AK167806.1 - 5 - 6 555 83
AK169066.1 - 5 - 6 551 83
AK169351.1 - 5 - 6 518 83
AK181035.1 - 2 - 3 172 83
AK181279.1 - 2 - 3 159 82
AK197714.1 - 2 - 3 172 83
AK207686.1 - 2 - 3 172 83
AK214436.1 - 2 - 3 172 83
BC119178.1 - 5 - 6 555 83
BC119180.1 - 5 - 6 555 83

View Full Product Details