Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lpo
Entrez Gene ID
76113
Gene Name
lactoperoxidase
Gene Aliases
5830499B15Rik
Chromosome Location
Chr.11: 87806428 - 87826135 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC016212.1 - 8 - 9 1479 89

View Full Product Details