The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028416.2 | NP_082692.1 | 3 - 4 | 103 | |
GenBank mRNA | AJ457192.1 | - | 3 - 4 | 103 | |
BC120532.1 | - | 3 - 4 | 374 | 103 | |
BC125438.1 | - | 3 - 4 | 374 | 103 |