Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Klhdc7b
Entrez Gene ID
546648
Gene Name
kelch domain containing 7B
Gene Aliases
EG546648, Gm306
Chromosome Location
Chr.15: 89386891 - 89388708 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160178.1 NP_001153650.1 1 - 1 1777 108

View Full Product Details