Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Asb8
Entrez Gene ID
78541
Gene Name
ankyrin repeat and SOCS box-containing 8
Gene Aliases
4930539L19Rik, AI788835, C430011H06Rik
Chromosome Location
Chr.15: 98134637 - 98165602 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170711.1 NP_001164182.1 1 - 2 105 60
GenBank mRNA AK016001.1 - 1 - 2 153 60

View Full Product Details