Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slfn5
Entrez Gene ID
327978
Gene Name
schlafen 5
Gene Aliases
AI956840
Chromosome Location
Chr.11: 82951349 - 82964850 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_183201.4 NP_899024.3 3 - 4 1189 90
GenBank mRNA AK036486.1 - 3 - 4 1190 90
AK040359.1 - 3 - 4 1176 90
AK171886.1 - 2 - 3 1131 90
AY261804.1 - 2 - 3 1151 90
AY261805.1 - 2 - 3 1146 90
BC117909.1 - 2 - 3 1069 90
BC117910.1 - 2 - 3 1069 90

View Full Product Details