Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cdh6
Entrez Gene ID
12563
Gene Name
cadherin 6
Gene Aliases
K-cadherin, cad6, cadherin-6
Chromosome Location
Chr.15: 13028699 - 13173689 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007666.3 NP_031692.2 3 - 4 781 54
GenBank mRNA AK134611.1 - 3 - 4 818 54
BC107008.1 - 2 - 3 522 54
BC107013.1 - 2 - 3 522 54
D82029.1 - 2 - 3 522 54

View Full Product Details