Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Chadl
Entrez Gene ID
214685
Gene Name
chondroadherin-like
Gene Aliases
AY100452, D930017K21Rik
Chromosome Location
Chr.15: 81686166 - 81697376 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164320.1 NP_001157792.1 5 - 6 2307 64
GenBank mRNA AK139481.1 - 2 - 3 871 64

View Full Product Details