The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026737.3 | NP_081013.1 | 3 - 4 | 271 | 82 |
GenBank mRNA | AF479288.1 | - | 3 - 4 | 287 | 82 |
AK003520.1 | - | 3 - 4 | 284 | 82 | |
AK143590.1 | - | 3 - 4 | 272 | 82 | |
AK163247.1 | - | 3 - 4 | 250 | 82 | |
AK167932.1 | - | 3 - 4 | 292 | 82 | |
AK167955.1 | - | 3 - 4 | 280 | 82 | |
BC025161.1 | - | 3 - 4 | 282 | 82 |