The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166456.1 | NP_001159928.1 | 10 - 11 | 1123 | 64 |
NM_001166458.1 | NP_001159930.1 | 10 - 11 | 1330 | 64 | |
NM_134086.4 | NP_598847.2 | 10 - 11 | 1816 | 64 | |
GenBank mRNA | AF184240.2 | - | 9 - 10 | 908 | 64 |
AK029189.1 | - | 10 - 11 | 993 | 64 | |
AK034130.1 | - | 10 - 11 | 1818 | 64 | |
AK035098.1 | - | 9 - 10 | 835 | 64 | |
AK047459.1 | - | 10 - 11 | 1347 | 64 | |
AK049173.1 | - | 10 - 11 | 1031 | 64 | |
AK049294.1 | - | 10 - 11 | 1096 | 64 | |
AK050914.1 | - | 10 - 11 | 1031 | 64 | |
AK081724.1 | - | 10 - 11 | 1332 | 64 | |
AK162612.1 | - | 10 - 11 | 1181 | 64 | |
AK163914.1 | - | 5 - 6 | 434 | 64 | |
AK165188.1 | - | 10 - 11 | 1383 | 64 | |
AK217621.1 | - | 3 - 4 | 194 | 64 | |
BC030378.1 | - | 10 - 11 | 1032 | 64 |