Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gucy1b2
Entrez Gene ID
239134
Gene Name
guanylate cyclase 1, soluble, beta 2
Gene Aliases
B930025B03, GC-SB2
Chromosome Location
Chr.14: 62373023 - 62457562 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204340.1 NP_001191269.1 5 - 6 666 72
NM_172810.3 NP_766398.1 5 - 6 666 72
GenBank mRNA AK047129.1 - 5 - 6 667 72
AY236513.1 - 3 - 4 340 72
BC137868.1 - 5 - 6 663 72
BC144988.1 - 5 - 6 663 72

View Full Product Details