The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178653.3 | NP_848768.1 | 3 - 4 | 509 | 102 |
GenBank mRNA | AK042919.1 | - | 3 - 4 | 397 | 102 |
AK046033.1 | - | 3 - 4 | 510 | 102 | |
AK046468.1 | - | 3 - 4 | 496 | 102 | |
AK050554.1 | - | 3 - 4 | 458 | 102 | |
AK077695.1 | - | 3 - 4 | 459 | 102 | |
AK082803.1 | - | 3 - 4 | 394 | 102 | |
AK086448.1 | - | 3 - 4 | 532 | 102 | |
AK086632.1 | - | 3 - 4 | 507 | 102 | |
AK145399.1 | - | 3 - 4 | 493 | 102 | |
AK166018.1 | - | 3 - 4 | 421 | 102 | |
AK215316.1 | - | 3 - 4 | 251 | 102 | |
BC025803.1 | - | 3 - 4 | 506 | 102 |