Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ttll7
Entrez Gene ID
70892
Gene Name
tubulin tyrosine ligase-like family, member 7
Gene Aliases
1110049N09Rik, 4921517B04Rik, C630030B20
Chromosome Location
Chr.3: 146852136 - 146984009 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK083236.1 - 5 - 6 710 81
AM690751.1 - 20 - 21 2615 81
BC138274.1 - 5 - 6 837 81
BC138275.1 - 5 - 6 837 81

View Full Product Details