Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mcoln2
Entrez Gene ID
68279
Gene Name
mucolipin 2
Gene Aliases
3300002C04Rik, AI549968, C86638, TRPML2
Chromosome Location
Chr.3: 146149796 - 146195514 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005846.2 NP_001005846.1 2 - 3 307 77
NM_026656.4 NP_080932.2 2 - 3 335 77
GenBank mRNA AF503575.1 - 2 - 3 324 77
AK014467.1 - 2 - 3 238 77
AK019454.1 - 2 - 3 204 77
AK150446.1 - 2 - 3 303 77
AK150547.1 - 2 - 3 303 77
AK152067.1 - 2 - 3 303 77
AK152943.1 - 2 - 3 303 77
AK153290.1 - 2 - 3 303 77
AK159172.1 - 2 - 3 335 77
AY083532.1 - 1 - 2 153 77
BC029847.1 - 2 - 3 184 77
CJ134616.1 - 2 - 3 307 77

View Full Product Details