Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bmpr1b
Entrez Gene ID
12167
Gene Name
bone morphogenetic protein receptor, type 1B
Gene Aliases
AI385617, ALK-6, AV355320, Acvrlk6, Alk6, BMPR-1B, BMPR-IB, CFK-43a, SKR6
Chromosome Location
Chr.3: 141837136 - 142169590 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277216.1 NP_001264145.1 4 - 5 417 99
NM_001277217.1 NP_001264146.1 4 - 5 443 99
NM_001277218.1 NP_001264147.1 3 - 4 456 99
NM_001277220.1 NP_001264149.1 4 - 5 591 99
NM_007560.4 NP_031586.1 3 - 4 610 99
GenBank mRNA AK086130.1 - 4 - 5 403 99
AK140158.1 - 4 - 5 515 99
AK162844.1 - 3 - 4 291 99
BC065106.1 - 3 - 4 391 99
BC065143.1 - 3 - 4 597 99
BC138638.1 - 3 - 4 337 99
BC138640.1 - 3 - 4 337 99
Z23143.1 - 4 - 5 435 99

View Full Product Details