Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rnf31
Entrez Gene ID
268749
Gene Name
ring finger protein 31
Gene Aliases
AL033293, BC031509, Flj10111, HOIP, Paul, mFLJ00217
Chromosome Location
Chr.14: 55591713 - 55603671 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_194346.2 NP_919327.2 16 - 17 2799 77
GenBank mRNA AK131160.1 - 9 - 10 4358 77
AK156320.1 - 16 - 17 2882 77
AY283085.1 - 16 - 17 2708 77
BC057595.1 - 16 - 17 2799 77

View Full Product Details