Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Diexf
Entrez Gene ID
215193
Gene Name
digestive organ expansion factor homolog (zebrafish)
Gene Aliases
AA408296, AW045375, Def
Chromosome Location
Chr.1: 193104403 - 193130285 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145415.2 NP_663390.2 3 - 4 529 81
GenBank mRNA AK052355.1 - 3 - 4 530 81
AK147251.1 - 3 - 4 523 81
BC019218.1 - 4 - 5 489 81

View Full Product Details