Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Esrrg
Entrez Gene ID
26381
Gene Name
estrogen-related receptor gamma
Gene Aliases
ERR3, Errg, NR3B3, mKIAA0832
Chromosome Location
Chr.1: 187608792 - 188214884 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243792.1 NP_001230721.1 7 - 8 1367 68
NM_011935.3 NP_036065.1 6 - 7 1305 68
GenBank mRNA AB093266.1 - 7 - 8 1386 68
AB221580.1 - 6 - 7 1139 68
AF117254.1 - 6 - 7 1202 68
AK052731.1 - 7 - 8 1346 68
AK140676.1 - 4 - 5 654 68
BC082324.1 - 6 - 7 1273 68

View Full Product Details