The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015772.3 | NP_056587.2 | 1 - 2 | 569 | 102 |
GenBank mRNA | AB093233.1 | - | 1 - 2 | 228 | 102 |
BC054527.1 | - | 1 - 2 | 264 | 102 | |
BC085361.1 | - | 1 - 2 | 346 | 102 | |
CB525158.1 | - | 1 - 2 | 351 | 102 |