The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252525.1 | NP_001239454.1 | 9 - 10 | 1527 | 87 |
NM_001252526.1 | NP_001239455.1 | 9 - 10 | 1509 | 87 | |
NM_007755.5 | NP_031781.1 | 9 - 10 | 1524 | 87 | |
GenBank mRNA | AK077799.1 | - | 1 - 2 | 152 | 87 |
AK136088.1 | - | 9 - 10 | 1528 | 87 | |
BC125476.1 | - | 9 - 10 | 1444 | 87 | |
BC144948.1 | - | 9 - 10 | 1426 | 87 |