The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291280.1 | NP_001278209.1 | 1 - 2 | 237 | 77 |
NM_026523.4 | NP_080799.1 | 1 - 2 | 237 | 77 | |
GenBank mRNA | AA671747.1 | - | 1 - 2 | 208 | 77 |
AK011929.1 | - | 1 - 2 | 194 | 77 | |
AK014077.1 | - | 1 - 2 | 216 | 77 | |
BC028490.1 | - | 1 - 2 | 172 | 77 | |
CB058776.1 | - | 1 - 2 | 249 | 77 |