Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Defb30
Entrez Gene ID
73670
Gene Name
defensin beta 30
Gene Aliases
2410125J01Rik, 4930449O14Rik, BD-30, mBD-30
Chromosome Location
Chr.14: 63034087 - 63037846 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK010780.1 - - 197 71
AK019622.1 - - 201 71

View Full Product Details