The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001285780.1 | NP_001272709.1 | 8 - 9 | 740 | 109 |
NM_025435.3 | NP_079711.1 | 7 - 8 | 662 | 109 | |
GenBank mRNA | AK005176.1 | - | 7 - 8 | 589 | 109 |
AK007440.1 | - | 7 - 8 | 568 | 109 | |
AK010958.1 | - | 7 - 8 | 603 | 109 | |
AK147003.1 | - | 8 - 9 | 643 | 109 | |
BC027113.1 | - | 6 - 7 | 1610 | 109 | |
BC042209.1 | - | 7 - 8 | 589 | 109 | |
BC054419.1 | - | 7 - 8 | 622 | 109 | |
BC116919.1 | - | 7 - 8 | 627 | 109 | |
BC116945.1 | - | 7 - 8 | 627 | 109 | |
CF949442.1 | - | 2 - 3 | 134 | 109 |