The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001004185.3 | NP_001004185.3 | 8 - 9 | 1650 | 60 |
GenBank mRNA | AK220273.1 | - | 8 - 9 | 1650 | 60 |
BC042749.2 | - | 9 - 10 | 1485 | 60 | |
BC050804.1 | - | 8 - 9 | 1113 | 60 |