Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hr
Entrez Gene ID
15460
Gene Name
hairless
Gene Aliases
ALUNC, AU, N, ba, bldy, hr, rh, rh-bmh, rhino
Chromosome Location
Chr.14: 70554056 - 70573548 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021877.3 NP_068677.2 10 - 11 2882 59
GenBank mRNA AB221596.1 - 5 - 6 567 59
AK076280.1 - 4 - 5 367 59
AY312855.1 - 9 - 10 2565 59
AY547390.1 - 9 - 10 2562 59
AY547391.1 - 9 - 10 2562 59
BC049182.1 - 10 - 11 2882 59
Z32675.1 - 9 - 10 2565 59

View Full Product Details