Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cdca2
Entrez Gene ID
108912
Gene Name
cell division cycle associated 2
Gene Aliases
2610311M19Rik, AI586158
Chromosome Location
Chr.14: 67673180 - 67715841 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110162.1 NP_001103632.1 8 - 9 1408 105
NM_175384.4 NP_780593.3 8 - 9 1111 105
GenBank mRNA AK031313.1 - 4 - 5 553 105
AK160223.1 - 8 - 9 1170 105
AK166952.1 - 8 - 9 1178 105
AK208630.1 - 1 - 2 68 105
AK208800.1 - 1 - 2 68 105
BC067009.1 - 8 - 9 1139 105
BC075682.1 - 8 - 9 1111 105
BC116298.1 - 4 - 5 553 105
BC116299.1 - 4 - 5 553 105

View Full Product Details