Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Npnt
Entrez Gene ID
114249
Gene Name
nephronectin
Gene Aliases
1110009H02Rik, AA682063, AI314031, Nctn, POEM
Chromosome Location
Chr.3: 132881745 - 132950291 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029836.2 NP_001025007.1 8 - 9 1408 125
NM_001287101.1 NP_001274030.1 10 - 11 1552 125
NM_001287102.1 NP_001274031.1 9 - 10 1501 125
NM_001287103.1 NP_001274032.1 8 - 9 1126 125
NM_033525.3 NP_277060.2 9 - 10 1459 125
GenBank mRNA AB059656.1 - 8 - 9 1408 125
AF397007.1 - 9 - 10 1253 125
AF397008.1 - 10 - 11 1304 125
AK050484.1 - 8 - 9 1312 125
AK158764.1 - 8 - 9 1128 125
AY035898.1 - 8 - 9 1283 125
AY035899.1 - 9 - 10 1334 125
BC046642.1 - 3 - 4 595 125
BC068308.1 - 8 - 9 1277 125

View Full Product Details