Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rwdd3
Entrez Gene ID
66568
Gene Name
RWD domain containing 3
Gene Aliases
2510027J23Rik, 3110037C01Rik, AI035684, AI662458, Rsume, X2CR1
Chromosome Location
Chr.3: 121155400 - 121171780 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025637.3 NP_079913.2 5 - 6 927 131
NM_028456.3 NP_082732.2 3 - 4 711 131
GenBank mRNA AF439556.1 - 3 - 4 710 131
AK014130.1 - 3 - 4 649 131
AK134288.1 - 3 - 4 702 131
BC037515.1 - 1 - 2 310 131
BC128287.1 - 3 - 4 710 131
BC128288.1 - 5 - 6 926 131
BC139340.1 - 3 - 4 710 131
BC139342.1 - 3 - 4 710 131

View Full Product Details