Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rwdd3
Entrez Gene ID
66568
Gene Name
RWD domain containing 3
Gene Aliases
2510027J23Rik, 3110037C01Rik, AI035684, AI662458, Rsume, X2CR1
Chromosome Location
Chr.3: 121155400 - 121171780 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025637.3 NP_079913.2 6 - 7 1037 142
NM_028456.3 NP_082732.2 4 - 5 821 142
GenBank mRNA AK014130.1 - 4 - 5 759 142
AK134288.1 - 4 - 5 812 142
BC128287.1 - 4 - 5 820 142
BC128288.1 - 6 - 7 1036 142

View Full Product Details