The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199465.2 | NP_955759.2 | 10 - 11 | 1420 | 66 |
GenBank mRNA | AK135919.1 | - | 1 - 2 | 108 | 66 |
AK162232.1 | - | 5 - 6 | 862 | 66 | |
AK162924.1 | - | 2 - 3 | 332 | 66 | |
BC052878.1 | - | 10 - 11 | 1367 | 66 | |
HQ179982.1 | - | 10 - 11 | 1454 | 66 | |
HQ179983.1 | - | 8 - 9 | 1223 | 66 | |
HQ179984.1 | - | 7 - 8 | 1052 | 66 |