The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025662.5 | NP_079938.1 | 9 - 10 | 1017 | 58 |
NM_178016.3 | NP_821135.1 | 10 - 11 | 1158 | 58 | |
GenBank mRNA | AK013853.1 | - | 9 - 10 | 1014 | 58 |
AK037552.1 | - | 5 - 6 | 601 | 58 | |
AK043033.1 | - | - | 1006 | 58 | |
AK046246.1 | - | 10 - 11 | 1146 | 58 | |
AK077893.1 | - | 9 - 10 | 1018 | 58 | |
AK082806.1 | - | 9 - 10 | 1010 | 58 | |
AK168666.1 | - | - | 999 | 58 | |
BC023256.1 | - | 9 - 10 | 999 | 58 | |
BC047203.1 | - | 5 - 6 | 555 | 58 | |
BC060175.1 | - | 9 - 10 | 1003 | 58 |