The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081094.1 | NP_001074563.1 | 1 - 2 | 783 | 61 |
GenBank mRNA | AK148729.1 | - | 1 - 2 | 760 | 61 |
AK167835.1 | - | 2 - 3 | 205 | 61 | |
AK168844.1 | - | 2 - 3 | 199 | 61 | |
BC147498.1 | - | 1 - 2 | 669 | 61 | |
BC150966.1 | - | 1 - 2 | 669 | 61 | |
BC150967.1 | - | 1 - 2 | 669 | 61 | |
BC151198.1 | - | 1 - 2 | 669 | 61 |