Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Gstcd
- Entrez Gene ID
-
67553
- Gene Name
- glutathione S-transferase, C-terminal domain containing
- Gene Aliases
-
4933434L15Rik
- Chromosome Location
-
Chr.3: 132981752 - 133091971 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK044441.1 |
- |
9 - 10 |
1845 |
71 |
View Full Product Details