Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zfand6
Entrez Gene ID
65098
Gene Name
zinc finger, AN1-type domain 6
Gene Aliases
3110005P07Rik, Awp1, Za20d3
Chromosome Location
Chr.7: 84615045 - 84689952 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AJ251508.1 - - 106 58
BC010683.1 - 1 - 2 61 58

View Full Product Details