The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019920.2 | NP_064304.1 | 4 - 5 | 250 | 100 |
GenBank mRNA | AF082526.1 | - | 4 - 5 | 250 | 100 |
AK088454.1 | - | 4 - 5 | 257 | 100 | |
AK151432.1 | - | 4 - 5 | 280 | 100 | |
AK159450.1 | - | 4 - 5 | 248 | 100 | |
AK162102.1 | - | 4 - 5 | 242 | 100 | |
AK211475.1 | - | 4 - 5 | 185 | 100 | |
BC016417.1 | - | 4 - 5 | 233 | 100 | |
BC083155.1 | - | 4 - 5 | 232 | 100 |