Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Klhl33
Entrez Gene ID
546611
Gene Name
kelch-like 33
Gene Aliases
EG546611
Chromosome Location
Chr.14: 50891336 - 50896531 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166651.1 NP_001160123.1 2 - 3 1051 85

View Full Product Details