Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ttc23
Entrez Gene ID
67009
Gene Name
tetratricopeptide repeat domain 23
Gene Aliases
1600012K10Rik
Chromosome Location
Chr.7: 67647410 - 67730060 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK086049.1 - - 1799 103

View Full Product Details