Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Smyd3
Entrez Gene ID
69726
Gene Name
SET and MYND domain containing 3
Gene Aliases
2410008A19Rik, Zmynd1
Chromosome Location
Chr.1: 178951960 - 179518052 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK046829.1 - 2 - 3 195 109

View Full Product Details