Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tacr3
Entrez Gene ID
21338
Gene Name
tachykinin receptor 3
Gene Aliases
Nk3r, Nkr, Tac3r
Chromosome Location
Chr.3: 134829007 - 134934581 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021382.6 NP_067357.1 1 - 2 773 93
GenBank mRNA AF233341.1 - 1 - 2 507 93
AK031898.1 - 1 - 2 774 93
AK046424.1 - 1 - 2 146 93
BC066845.1 - 1 - 2 744 93
L27827.1 - 1 - 2 201 93
X87823.1 - 1 - 2 731 93

View Full Product Details